missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ VDAC3 Recombinant Protein

Brand:  Abnova™ H00007419-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16121822

  • 6845.00 SEK / 25µg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH56870
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
56.87
8
Glutathione Sepharose 4 Fast Flow
25 ug
MCNTPTYCDLGKAAKDVFNKGYGFGMVKIDLKTKSCSGVEFSTSGHAYTDTGKASGNLETKYKVCNYGLTFTQKWNTDNTLGTEISWENKLAEGLKLTLDTIFVPNTGKKSGKLKASYKRDCFSVGSNVDIDFSGPTIYGWAVLAFEGWLAGYQMSFDTAKSKLSQNNFALGYKAADFQLHTHVNDGTEFGGSIYQKVNEKIETSINLAWTAGSNNTRFGIAAKYMLDCRTSLSAKVNNASLIGLGYTQTLRPGVKLTLSALIDGKNFSAGGHKVGLGFELEA
HD-VDAC3
VDAC3
Wheat Germ (in vitro)
GST
Antibody Production, ELISA, Protein Array, Western Blot
7419
VDAC3 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
VDAC3
Human
Recombinant
Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers