Läs mer
Venlafaxine EP Impurity B (as Hydrochloride), Mikromol™
Beskrivning
Venlafaxine EP Impurity B (as Hydrochloride)
Specifikationer
Specifikationer
| Kemiskt namn eller material | Ethyl (2RS)-3-(Dimethylamino)-2-(4-methoxyphenyl)propanoate Hydrochloride |
| CAS | 2174001-92-2 |
| Analyt- eller komponentnamn | Ethyl (2RS)-3-(Dimethylamino)-2-(4-methoxyphenyl)propanoate Hydrochloride |
| Frakt skick | Room Temperature |
| Molekylformel | C14 H21 N O3 . Cl H |
| Certifikat och efterlevnad | ISO 17025 |
| Synonym | Venlafaxine Hydrochloride Imp. B (EP), Venlafaxine Imp. B (EP), Ethyl (2RS)-3-(Dimethylamino)-2-(4-methoxyphenyl)propanoate Hydrochloride, Venlafaxine Hydrochloride Imp. B (EP) as Hydrochloride, Venlafaxine Hydrochloride Impurity B as Hydrochloride, Venlafaxine Impurity B as Hydrochloride |
| InChI formel | InChI=1S/C14H21NO3.ClH/c1-5-18-14(16)13(10-15(2)3)11-6-8-12(17-4)9-7-11;/h6-9,13H,5,10H2,1-4H3;1H |
| LEDER | Cl.CCOC(=O)C(CN(C)C)c1ccc(OC)cc1 |
| IUPAC-namn | ethyl 3-(dimethylamino)-2-(4-methoxyphenyl)propanoate;hydrochloride |
| Visa mer |