Learn More
Veolia Water Systems™ UV Lamp
Rich, robust UV lamp that features UV light acting as a bactericide to break down and photo-oxidize organic contaminants to polar or ionized species.
Brand:  Veolia Water Systems LC118
Additional Details : Weight : 0.09180kg
 View more versions of this product
Product Code. 11974897
  • 7950.00 SEK / Each
Estimated Shipment: 11-04-2024
to see stock.
Add to basket
Description
Description
  • UV light is used as a bactericide and to break down and photo-oxidise organic contaminants to polar or ionised species
  • The UV bactericidal action damages DNA and RNA polymerase at low doses thereby preventing replication
Specifications
Specifications
Break down and photo-oxidize organic contaminants to polar or ionized species
Centra R 60/120, Maxima MK1/MK2, PureLab Option-Q, PureLab Ultra MK1, PureLab Ultra MK2