Läs mer
Vercirnon, MedChemExpress
Produktkod. 16492847
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16492847 50 mg
16489317 5 mg
16446896 100 mg
16406778 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16492847 Leverantör Medchem Express Leverantörsnummer HY15724
MedChemExpress Vercirnon (GSK1605786A) is an orally bioavailable, selective, and potent antagonist of CCR9.
Vercirnon inhibits CCR9-mediated Ca2+ mobilization and chemotaxis on Molt-4 cells with IC50 values of 5.4 and 3.4 nM, respectively. Vercirnon is selective for CCR9 over CCR1-12 and CX3CR1-7 (IC50s>10 μM for all). Vercirnon is an equipotent inhibitor of CCL25-directed chemotaxis of both splice forms of CCR9 (CCR9A and CCR9B) with IC50 values of 2.8 and 2.6 nM, respectively.
Specifikationer
Kemiskt namn eller material Vercirnon
CAS 698394-73-9
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C22H21ClN2O4S
Kvantitet 50 mg
Synonym GSK-1605786 CCX282-B Traficet-EN
Löslighetsinformation DMSO : ≥ 25 mg/mL (56.19 mM)
LEDER O=S(C1=CC=C(C(C)(C)C)C=C1)(NC2=CC=C(Cl)C=C2C(C3=CC=[N+]([O-])C=C3)=O)=O
Molekylvikt (g/mol) 444.93
Formel vikt 444.93
Procent renhet 98.19%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) COVID-19-immunoregulation