Läs mer
Tocris Bioscience™ Verdinexor
Beskrivning
Verdinexor is a selective exportin-1 (XPO1/CRM1) inhibitor. It potently inhibits influenza virus ribonucleoprotein (vRNP) export by disrupting XPO1-NEP binding and the replication of influenza virus A and B strains in vitro, including H1N1, H5N1 and H7N9 (IC50 values in the range 40 - 420 nM range). Verdinexor limits influenza A virus spread in mouse lung in vivo. In neuroblastoma, verdinexor suppresses cell proliferation and induces cell apoptosis. It also induces P53 nuclear accumulation and induces G0/G1 phase cell cycle arrest by activating P53 function. Verdinexor inhibits tumor growth in in vivo mouse models. Orally bioavailable.
Specifikationer
Specifikationer
| Kemiskt namn eller material | (2 Z)-3-[3-[3,5-Bis(trifluoromethyl)phenyl]-1 H-1,2,4-triazol-1-yl]-2-propenoic acid 2-(2-pyridinyl)hydrazide |
| CAS | 1392136-43-4 |
| Kvantitet | 10 mg |
| Mål | Antiviral Compounds |
| Molekylformel | C18H12F6N6O |
| Renhet | 0.98 |