Läs mer
Phenomenex™ Verex™ Preassembled Screw Caps, 9 mm, with Septa, No Slit
Beskrivning
- Analysis: Cleanliness and performance tested as standard
- Certification: 100% defect-free
- Configurations: Specialty vials, crimp-top vials, screw-top vials, snap-top vials, VOA/ASE assembled vial kits, VOA/ASE assembled storage kits
Specifikationer
Specifikationer
| Färg | Blue |
| Diameter (metrisk) Yttre | 9 mm |
| Förslits | No |
| Sterilitet | Sterile |
| Septum | PTFE/Rubber |
| Kvantitet | 100 Pk. |
| Stängningsstorlek | 9 mm |
| Markeringspunkt | No |
| Autoklaveringsbar | Autoclavable |
| Produkttyp | Cap |