Läs mer
Phenomenex™ Verex™ Vial Inserts, 5 mm, Conical Bottom, Glass
Produktkod. 31748383 Handla allt Phenomenex Produkter
Change view
Kvantitet:
100 Pk.
1000 Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
31748383 100 Pk.
31742947 1000 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 31748383 Leverantör Phenomenex™ Leverantörsnummer AR0452012
Beräknad leverans: 21-09-2026
Accessories for use with chromatography vials and in various chromatography applications.
  • Analysis: Cleanliness and performance tested as standard
  • Certification: 100% defect-free
  • Configurations: Specialty vials, crimp-top vials, screw-top vials, snap-top vials, VOA/ASE assembled vial kits, VOA/ASE assembled storage kits
TRUSTED_SUSTAINABILITY
Specifikationer
Färg Clear
Diameter (metrisk) Yttre 12 mm
Material Glass
Förslits No
Sterilitet Sterile
Kvantitet 100 Pk.
Stängningsstorlek 5 mm
Markeringspunkt No
Autoklaveringsbar Autoclavable
Produkttyp Insert