Läs mer
Verruculogen, TRC
Produktkod. 30454888
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30454888 1 mg
1 options
Produktkod. 30454888 Leverantör Toronto Research Chemicals Leverantörsnummer TRCV2671901MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Verruculogen
CAS 12771-72-1
Molekylformel C27H33N3O7
Kvantitet 1 mg
Synonym 5H,12H-3,4-Dioxa-5a,11a,15a-triazacyclooct[lm]indeno[5,6-b]fluorene-11,15(2H,13H)-dione','1,10,10a,14,14a,15b-hexahydro-10,10a-dihydroxy-7-methoxy-2,2-dimethyl-5-(2-methyl-1-propenyl)-','(5R,10S,10aR,14aS,15bS)- (9CI), 5H,12H-3,4-Dioxa-5a,11a,15a-triazacyclooct[lm]indeno[5,6-b]fluorene-11,15(2H,13H)-dione','1,10,10a,14,14a,15b-hexahydro-10,10a-dihydroxy-7-methoxy-2,2-dimethyl-5-(2-methyl-1-propenyl)-','[5R-(5α,10α,10aα,14aα,15bα)]-, (5R,10S,10aR,14aS,15bS)-1,10,10a,14,14a,15b-Hexahydro-10,10a-dihydroxy-7-methoxy-2,2-dimethyl-5-(2-methyl-1-propen-1-yl)-5H,12H-3,4-dioxa-5a,11a,15a-triazacyclooct[lm]indeno[5,6-b]fluorene-11,15(2H,13H)-dione, NA 209A, TR 1, TR 1 toxin, Verruculogen, Verruculogen TR 1
InChI formel InChI=1S/C27H33N3O7/c1-14(2)11-20-29-18-12-15(35-5)8-9-16(18)21-22(29)19(13-26(3,4)37-36-20)30-24(32)17-7-6-10-28(17)25(33)27(30,34)23(21)31/h8-9,11-12,17,19-20,23,31,34H,6-7,10,13H2,1-5H3/t17-,19-,20+,23-,27+/m0/s1
LEDER COc1ccc2c3[C@H](O)[C@]4(O)N([C@H]5CC(C)(C)OO[C@H](C=C(C)C)n(c35)c2c1)C(=O)[C@@H]6CCCN6C4=O
Molekylvikt (g/mol) 511.57
Formel vikt 511.2319