missing translation for 'onlineSavingsMsg'
Learn More

3M™ Versaflo™ Filter Cover TR-371+

3M™ Versaflo™ Filter Cover TR-371+ for use with Versaflo™ TR-300+ Powered Air Turbo.

Brand:  3M™ 7100099577

202.44 SEK valid until 2023-12-31
Use promo code "20707" to get your promotional price.



Additional Details : Weight : 0.50000kg

Product Code. 15608576

  • 253.62 SEK / Each

Estimated Shipment: 18-10-2023
to see stock.
Add to basket

Description

Description

  • Holds TR-3712E, TR-3802E, and TR-3822E filters on turbo unit
  • Quick view window enables users to easily see filter label
  • Easy to install
Specifications

Specifications

CE Approved
Filter Cover
0.21 x 0.132 x 0.076 m
Holds TR-3712E, TR-3802E and TR-3822E filters on turbo unit
Versaflo™ TR-300+ Powered Air turbo
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers