Learn More
3M™ Versaflo™ S-Series Painter’s Hood Assembly with Inner Shroud and Helmet, S-767
Product Code. 31297046 Shop All 3M Products
Change view
Click to view available options
Product Type:
PAPR Assembly with Hood
Product Code. Product Type
31297046 PAPR Assembly with Hood
1 options
Product Code. 31297046 Supplier 3M™ Supplier No. 7100325969
  • ONLINE EXCLUSIVE PRICE
    3545.00 SEK
    2210.39 SEK / Paket med 1
    Save 1334.61 SEK
    37% Av
 Restricted Item
Beräknad leverans: 01-12-2026
Offer a variety of options for faceseals, collars, shrouds, and suspensions, allowing you to choose the respiratory protection that provides the most value and comfort for your team.
  • Lightweight, versatile and easy to maintain
  • Help protect your workers from a variety of contaminants while maintaining comfort
  • Available in fully assembled integrated hoods for convenience
  • Reusable suspension options offer cost savings
  • Designed to work for your needs
TRUSTED_SUSTAINABILITY
Specifications
Color White
Disposable Single-use
Filter Type Combination
Material Plastic
Product Type PAPR Assembly with Hood
Size One Size
With Exhalation Valve No