Läs mer
Thermo Scientific™ Versette™ Pipetting Heads
Produktkod. p-4528657
Kanaler:
96
384
Volym (metrisk):
0,5 till 30 μl
1 till 100 μl
5 till 300 μl
Förpackningsstorlek:
Varje
Produktkod. 15473770
Brand: Thermo Scientific™ 65006384100
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Choose from a volume range of 0.5μL to 300μL with Thermo Scientific™ Versette™ Pipetting Heads.
Compatible with multiple interchangeable pipetting heads, the Thermo Scientific™ Versette™ Automated Liquid Handler provides versatile pipetting options for a broad range of applications that require 96– to 384–channel automated pipetting. The Versette system is compatible with disposable D.A.R.T.'S™, with a total volume ranging from 0.5–300μl.
The 96– and 384–channel pipetting heads use Thermo Scientific D.A.R.T.'S™, which ensure effective sealing that provides accurate and precise pipetting across all channels.
  • All pipetting heads utilize RFID tags for self-identity
TRUSTED_SUSTAINABILITY
Specifikationer
Noggrannhet 2.0% or 0.5µL
Produkttyp Pipetting Head
Beskrivning 384 channels
Kanaler 384
Volym (metrisk) 1 till 100 μl
Precision 1.5% or 0.25 µL