Learn More
Systec™ Systec V-100/VX-1100 Vertical Floor-Standing Top-Loading Autoclave
Top-loading vertical autoclave with a small footprint and high chamber capacity. Systec™ Systec V-100/VX-1100 Vertical Floor-Standing Top-Loading Autoclave features state-of-the-art sterilization process capacities and options.
Brand: Systec™ 1100
Description
- Integrated, separate steam generator
- Housing, support frame and pressure vessel made of corrosion-resistant stainless steel
- LCD display and fully automatic microprocessor control
- Code-secured access for changing parameters and safety-relevant intervention
- Internal memory for storing up to 500 sterilization cycles
- Timer for starting programs
- Autofill: automatic demineralized water feed for steam generation
- Flexible PT-100 temperature sensor
- Additional temperature sensor in condense exhaust
- Temperature holding function for liquids after program finish
- Special program for Durham tubes
- Calculation of F0 value
- Special program for waste sterilization with pulsed heat-up for more efficient air exhaust
- Water-cooled steam exhaust, thermostatically controlled
- Programmable automatic door-opening on completion of program
- RS-232 and RS-485 interfaces for external data transmission (network-compatible)
- Additional options for control, process optimization, and documentation available
Specifications
117 L (Total), 100 L (Nominal) | |
500 mm | |
3 x 22 250mL Erlenmeyer flasks, 3 x 30 250mL Schott Duran™ media bottles | |
175 kg | |
RS-232 / RS-485 (Ethernet network and USB compatible) | |
900 x 650 x 985 mm Exterior |
500 mm | |
VX-100 | |
140°C | |
380 to 400V, 50/ 60 Hz, 16 A. Other voltage supplies available on request. Country-specific deviations possible. | |
4 bar max. |