Learn More
Scientific Industries SI™ Vertical Microtube Holder
Requires no additional attachment for use. Scientific Industries SI™ Vertical Microtube Holder snaps microtubes into place.
Brand: Scientific Industries SI™ 9730128
Additional Details : Weight : 1.00000kg
Description
- Holds tubes vertically
- Snaps on to any Vortex-Genie 2 Mixer and the Vortex-Genie Pulse
- Optimized for high speed vortexing a small number of tubes
- No additional attachment necessary
Specifications
Vortex-Genie 2 Mixer and Vortex-Genie Pulse | |
9 |