Learn More
Scientific Industries SI™ Vertical Microtube Holder
Requires no additional attachment for use. Scientific Industries SI™ Vertical Microtube Holder snaps microtubes into place.
Brand:  Scientific Industries SI™ 9730128
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 15196812
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Holds tubes vertically
  • Snaps on to any Vortex-Genie 2 Mixer and the Vortex-Genie Pulse
  • Optimized for high speed vortexing a small number of tubes
  • No additional attachment necessary
Specifications
Specifications
Vortex-Genie 2 Mixer and Vortex-Genie Pulse
9