Läs mer
Vesatolimod, MedChemExpress
Produktkod. 16486668
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
Förpackningsstorlek:
10mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16486668 10 mg 10mg
16469227 5 mg -
16436826 100 mg -
16402757 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16486668 Leverantör Medchem Express Leverantörsnummer HY15601
MedChemExpress Vesatolimod (GS-9620) is a potent, selective and orally active agonist of Toll-Like Receptor (TLR7) with an EC50 of 291 nM.
Vesatolimod (GS-9620) is a potent, selective and orally active agonist of Toll-Like Receptor (TLR7) with an EC50 of 291 nM.
Specifikationer
Kemiskt namn eller material Vesatolimod
Fysisk form Solid
CAS 1228585-88-3
Färg White
Kvantitet 10 mg
Anteckningar om renhetsgrad Research
Molekylformel C22H30N6O2
För användning med (applikation) COVID-19-anti-virus
Synonym GS-9620
Löslighetsinformation DMSO : ≥ 16.67 mg/mL (40.61 mM)
LEDER O=C1NC2=C(N)N=C(OCCCC)N=C2N(CC3=CC=CC(CN4CCCC4)=C3)C1
Molekylvikt (g/mol) 410.51
Formel vikt 410.51
Procent renhet 95.11%