Learn More
Vezatin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA552115
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: DELEKLVCTK ETQELVSEAY PILEQKLKLI QPHVQASNNC WEEAISQVDK LLRRNTDKKG KPEIACENPH CTVVPLKQPT LHIADKDPIP EEQELEAYVD DIDIDSDFRK DDFYYLSQED KERQKREHEE SKRVLQELKS VLG Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 83%.
Vezatin Polyclonal antibody specifically detects Vezatin in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
Vezatin | |
Polyclonal | |
Unconjugated | |
VEZT | |
A630071D13Rik; AI848282; transmembrane protein vezatin; vezatin; vezatin, adherens junctions transmembrane protein; VEZT | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55591 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9HBM0 | |
VEZT | |
Recombinant protein corresponding to Human Vezatin | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only