missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEZT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17684-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Plays a pivotal role in the establishment of adherens junctions and their maintece in adult life. In case of Listeria infection, promotes bacterial internalization by participating to myosin VIIa recruitment to the entry site.Specifications
VEZT | |
Polyclonal | |
Unconjugated | |
VEZT | |
VEZATIN, VEZT | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
215008, 299738, 55591 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q3ZK22, Q5XI52, Q9HBM0 | |
VEZT | |
VEZT Fusion Protein Ag11786 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |