Learn More
3M™ VFlex™ Particulate Respirato, 9163E
3M™ VFlex™ Particulate Respirator 9163E provides economic FFP3 protection and has a compliable V-shaped pleat design.
Brand: 3M™ 7100181483
Additional Details : Weight : 0.50000kg
Description
- Economic respirator solution
- V-shaped pleats flex and expand around your mouth to aid comfort
- Proprietary 3M™ filter material helps you breathe easier
- Embossed front panel retains shape and keeps material away from the mouth
- Side tabs enable easier positioning
- Embedded, adjustable metal nose clip helps provide a custom and secure seal
- Flat-fold design makes it convenient and easy to store away
- Designed to work well with 3M™ Eyewear and Hearing Protection products
- Available in small and regular sizes to fit different face shapes
- 3M™ Cool Flow™ Valve helps improve breathing comfort
Specifications
Particulate Respirator | |
9163E |