Learn More
VGLL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA559968
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: LSNIKSPQEL TPSSQSEGVM LKNDDSMSPN QWRYSSPWTK PQPEVPVTNR AANCNLHVPG PMAVNQFSPS LARRASVRPG ELWHFSSLAG TSSLEPGYSH PFPARHLVPE PQPDGKRE Highest antigen sequence identity to the following orthologs: Mouse - 33%, Rat - 39%.
May act as a specific coactivator for the mammalian TEFs.Specifications
VGLL1 | |
Polyclonal | |
Unconjugated | |
VGLL1 | |
Protein TONDU; RGD1564038; TDU; TONDU; Transcription cofactor vestigial-like protein 1; vestigial like 1; vestigial like 1 homolog; vestigial like 1 homolog (Drosophila); vestigial like family member 1; vestigial-like family member 1; vestigial-related factor; VGL1; Vgl-1; VGLL1; WUGSC:H_GS188P18.1b | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51442 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q99990 | |
VGLL1 | |
Recombinant protein corresponding to Human VGLL1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only