Läs mer
Tocris Bioscience™ VH 032 amide-PEG2-amine
Beskrivning
VH 032 amide-PEG2-amine is a functionalized von-Hippel-Lindau (VHL) protein ligand for PROTAC® research and development; incorporates an E3 ligase ligand plus a PEG linker ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.
This product has been recently renamed. The previous name for this product was VH 032 - linker 12
Please contact us for SD files of our available Degrader Building Blocks.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifikationer
Specifikationer
| Kemiskt namn eller material | (2S,4R)-1-((S)-2-(2-(2-(2-Aminoethoxy)ethoxy)acetamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide |
| CAS | 2341796-76-5 |
| Synonym | VH 032 - linker 12 |
| Kvantitet | 25 mg |
| Molekylformel | C28H41N5O6S.2HCl |
| Formel vikt | 648.64 |
| Produkttyp | Functionalized von-Hippel-Lindau (VHL) Ligand |
| Renhet | 95% |