Läs mer
VH 032, amine
Functionalized VHL ligand for PROTACs
Brand: Tocris Bioscience 6462/100
Beskrivning
VH 032, amine is a derivative of the von Hippel-Lindau (VHL) ligand, VH 032; commonly used as a precursor to a PROTAC® that hijacks VHL as the E3 ubiquitin ligase component. Supplied with a primary amine functional handle at a position known not to significantly affect binding to VHL, for ready conjugation to a linker/target protein ligand.PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Require a different derivative? Get in touch
Specifikationer
| (4 R)-3-Methyl-L-valyl-4-hydroxy-N-[[4-(4-methyl-5-thiazolyl)phenyl]methyl]-L-prolinamide dihydrochloride | |
| 2137142-47-1 | |
| (4R)-3-Methyl-L-valyl-4-hydroxy-N-[[4-(4-methyl-5-thiazolyl)phenyl]methyl]-L-prolinamide hydrochloride | |
| 100 mg |