Läs mer
VH 032 amide-PEG4-amine
VHL ligand with PEG linker and terminal amine for onward chemistry
Brand: Tocris Bioscience 6464/25
Beskrivning
VH 032 amide-PEG4-amine is a functionalized von-Hippel-Lindau protein ligand (VHL) for PROTAC® research and development; incorporates an E3 ligase ligand plus a PEG linker ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.This product has been recently renamed. The previous name for this product was VH 032 - linker 2
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifikationer
| (2 S,4 R)-1-(-17-amino-2-(tert-butyl)-4-oxo-6,9,12,15-tetraoxa-3-azaheptadecan-1-oyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide dihydrochloride | |
| 2446474-04-8 | |
| VH 032 - linker 2 | |
| 25 mg |