Läs mer
Tocris Bioscience™ VH 101, acid
Beskrivning
VH 101, acid is a functionalized von-Hippel-Lindau protein ligand (VHL) for PROTAC® research and development; incorporates an E3 ligase ligand with carboxylic acid handle at the benzylic position ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.
Please contact us for SD files of our available Degrader Building Blocks.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifikationer
Specifikationer
| Kemiskt namn eller material | -3-[[[(2 S,4 R)-1-[-2-(1-Fluorocyclopropan-1-ylcarbonylamino)-3,3-dimethylbutanoyl]-4-hydroxypyrrolidin-2-yl]carbonyl]amino]-3-[4-(4-methylthiazol-5-yl)phenyl]propanoic acid |
| CAS | 2408341-97-7 |
| Kvantitet | 50 mg |
| Mål | Degrader Building Blocks |
| Molekylformel | C28H35FN4O6S |
| Renhet | 0.98 |