Läs mer
Eurofins DiscoverX VHR Protein
Beskrivning
VHR protein is a member of the dual specificity protein phosphatase subfamily. These phosphatases inactivate their target kinases by dephosphorylating both the phosphoserine/threonine and phosphotyrosine residues. They negatively regulate members of the mitogen-activated protein (MAP) kinase superfamily (MAPK/ERK, SAPK/JNK, p38), which are associated with cellular proliferation and differentiation. Different members of the family of dual specificity phosphatases show distinct substrate specificities for various MAP kinases, different tissue distribution and subcellular localization, and different modes of inducibility of their expression by extracellular stimuli. This gene maps in a region that contains the BRCA1 locus which confers susceptibility to breast and ovarian cancer. Although DUSP3 is expressed in both breast and ovarian tissues, mutation screening in breast cancer pedigrees and in sporadic tumors was negative, leading to the conclusion that this gene is not BRCA1.
GenBank NM_004090
Specifikationer
Specifikationer
| Formulering | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 10% glycerol, 0.1 mM EGTA, 0.03% Brij-35, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Molekylvikt (g/mol) | 47.8 kDa |
| Kvantitet | 10 μg |
| Art | Human |
| Rekombinant | Recombinant |
| Protein Tag | N-terminal GST |
| Uttryckssystem | Recombinant enzyme expressed in E. coli cells |
| Proteinform | Full-length |
| Renhet eller kvalitetsklass | ≥78% |
| Protein | VHR, active |
Fisher Scientific Encompass-programmet erbjuder artiklar som inte ingår i vår distributionsportfölj. Dessa produkter har vanligtvis inga bilder eller detaljerade beskrivningar. Vi är dock angelägna om att förbättra din shoppingupplevelse. Använd formuläret nedan för att ge feedback relaterad till innehållet på denna produkt.