Learn More
Heidolph instruments Vial PolyBlock
Accommodate specifically sized vessels for your StarFish magnetic stirrer workstation. Heidolph™ Vial PolyBlock, is one of up to 5 StarFish PolyBlock segments that can be mixed at a single stirrer workstation to create various arrays of vessel size and type.
Brand: Heidolph instruments 505-80029-00
Additional Details : Weight : 0.32400kg
Description
- Accommodates one specific vial size.
- One of up to 5 segments per StarFish array.
Specifications
PolyBlock | |
Starfish Workstation, MR-Hei Magnetic Stirrers | |
9 x 12 mm Vials |