Läs mer
Viaminate, TRC
Produktkod. 30449103
Change view
Kvantitet:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30449103 250 mg
30452990 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30449103 Leverantör Toronto Research Chemicals Leverantörsnummer TRCV200120250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 53839-71-7
Molekylformel C29H37NO3
Molekylvikt (g/mol) 447.61
Synonym Viaminate, Retinamide, N-[4-(ethoxycarbonyl)phenyl]-, N-[4-(Ethoxycarbonyl)phenyl]retinamide, N-(p-Ethoxycarbonylphenyl)retinamide, Viaminate, N-[4-(Ethoxycarbonyl)phenyl]retinamide, N-(p-Ethoxycarbonylphenyl)retinamide, N-[4-(ethoxycarbonyl)phenyl]-Retinamide
IUPAC-namn ethyl 4-[[(2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenoyl]amino]benzoate
LEDER CCOC(=O)c1ccc(NC(=O)\C=C(/C)\C=C\C=C(/C)\C=C\C2=C(C)CCCC2(C)C)cc1
Specifikationer
Kemiskt namn eller material Viaminate
Kvantitet 250 mg
InChI formel InChI=1S/C29H37NO3/c1-7-33-28(32)24-14-16-25(17-15-24)30-27(31)20-22(3)11-8-10-21(2)13-18-26-23(4)12-9-19-29(26,5)6/h8,10-11,13-18,20H,7,9,12,19H2,1-6H3,(H,30,31)/b11-8+,18-13+,21-10+,22-20+
Formel vikt 447.2773