Läs mer
Vidarabine, TRC
Produktkod. 30454252
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30454252 500 mg
1 options
Produktkod. 30454252 Leverantör Toronto Research Chemicals Leverantörsnummer TRCV305030500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Vidarabine
CAS 5536-17-4
Molekylformel C10 H13 N5 O4
Kvantitet 500 mg
Synonym 9H-Purin-6-amine','9-β-D-arabinofuranosyl-, Adenine','9-β-D-arabinofuranosyl- (7CI,8CI), 9-β-D-Arabinofuranosyl-9H-purin-6-amine, 6-Amino-9-β-D-arabinofuranosylpurine, 9-Arabinosyladenine, 9-β-D-Arabinoadenosine, 9-β-D-Arabinofuranosyladenine, 9-β-D-Arabinosyladenine, Adenine 9-β-D-arabinofuranoside, Adenine arabinoside, Adenine β-D-arabinofuranoside, Ara-A, Araadenosine, Arabinosyladenine, Arasena-A, CI-673, NSC 247519, NSC 404241, Spongoadenosine, Vidarabin, Vidarabine, Vidarabine anhydrous, Vira-A, β-Ara-A, β-D-Arabinofuranosyladenine, β-D-Arabinosyladenine
InChI formel InChI=1S/C10H13N5O4/c11-8-5-9(13-2-12-8)15(3-14-5)10-7(18)6(17)4(1-16)19-10/h2-4,6-7,10,16-18H,1H2,(H2,11,12,13)/t4-,6-,7+,10-/m1/s1
LEDER Nc1ncnc2c1ncn2[C@@H]3O[C@H](CO)[C@@H](O)[C@@H]3O
IUPAC-namn (2R,3S,4S,5R)-2-(6-aminopurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
Molekylvikt (g/mol) 267.24
Formel vikt 267.0968