missing translation for 'onlineSavingsMsg'
Learn More

Vidarabine, TRC

Product Code. 30453083
Change view
Click to view available options
Kvantitet:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30453083 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30453083 Supplier Toronto Research Chemicals Supplier No. TRCV3050301G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Vidarabine
CAS 5536-17-4
Molekylformel C10 H13 N5 O4
Kvantitet 1 g
Synonym 9H-Purin-6-amine','9-β-D-arabinofuranosyl-, Adenine','9-β-D-arabinofuranosyl- (7CI,8CI), 9-β-D-Arabinofuranosyl-9H-purin-6-amine, 6-Amino-9-β-D-arabinofuranosylpurine, 9-Arabinosyladenine, 9-β-D-Arabinoadenosine, 9-β-D-Arabinofuranosyladenine, 9-β-D-Arabinosyladenine, Adenine 9-β-D-arabinofuranoside, Adenine arabinoside, Adenine β-D-arabinofuranoside, Ara-A, Araadenosine, Arabinosyladenine, Arasena-A, CI-673, NSC 247519, NSC 404241, Spongoadenosine, Vidarabin, Vidarabine, Vidarabine anhydrous, Vira-A, β-Ara-A, β-D-Arabinofuranosyladenine, β-D-Arabinosyladenine
InChI formel InChI=1S/C10H13N5O4/c11-8-5-9(13-2-12-8)15(3-14-5)10-7(18)6(17)4(1-16)19-10/h2-4,6-7,10,16-18H,1H2,(H2,11,12,13)/t4-,6-,7+,10-/m1/s1
LEDER Nc1ncnc2c1ncn2[C@@H]3O[C@H](CO)[C@@H](O)[C@@H]3O
IUPAC-namn (2R,3S,4S,5R)-2-(6-aminopurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
Molekylvikt (g/mol) 267.24
Formel vikt 267.0968
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.