Läs mer
Eutech™ Precision pH-millivolt Simulator, Hi-Low impedence
Produktkod. 11871253
Klicka för att se tillgängliga alternativ
Räckvidd (mV):
±2
Förpackningsstorlek:
Varje
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11871253
Brand: Thermo Scientific Eutech 01X373301
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Denna artikel kan inte returneras. Se returpolicy
The Eutech pH-millivolt simulator is a handy, easy-to-use service tool for testing the accuracy of your pH meter.
The Eutech pH-millivolt Simulator is a handy, easy-to-use service tool for testing the accuracy of your pH meter. Used for trouble-shooting and fault diagnosis of meters, the simulator works with most pH/redox instruments with BNC connectors. Simulator comes with a BNC cable and a sturdy rubber boot which doubles up as a stand.
Features
  • Push button pH/mV selection
  • pH accuracy of ±0.05; mV range of ±2
  • USA/NIST pH buffer standards simulation
  • 16 combination values
  • 1 GΩ high input impedance test
  • Large display with bright LED
  • Splashproof keypad
  • Protective rubber boot doubles up as stand
  • Power-saving meter runs on 4 AAA x 1.4 V batteries
  • TRUSTED_SUSTAINABILITY
    Specifikationer
    Noggrannhet (pH) ±0,05
    Visningstyp LED
    Mätlägen pH, Meter, Accuracy, Test, mV
    Räckvidd (mV) ±2
    Strömförsörjning 4 - AAA x 1.4V batteries