Läs mer
Invitrogen™ HMOX1 Monoclonal Antibody (HO-1-1)
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA1112
Produktkod. 13491007
  • 5380.00 SEK / 200µg
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
Denna artikel kan inte returneras. Se returpolicy
Beskrivning
Beskrivning
This antibody detects an ~32 kDa protein corresponding to the apparent molecular mass of HO-1 on SDS-PAGE immunoblots, in samples from human, mouse, rat, bovine and dog origins. This antibody does not cross-react with HO-2. This antibody was originally validated as part of a Thermo Scientific Cellomics High Content Screening Kit. The antibody sold separately may have slightly different performance and may need to be further optimized for the best results.
Heme oxygenase (HO) is a microsomal enzyme that catalyzes the oxidation of heme to the antioxidant molecules, biliverdin and carbon monoxide. HO consists of two homologous isozymes, an inducible HO-1 and a constitutively expressed HO-2. HO-1 is induced by a wide variety of stimuli including conditions of oxidative stress, inflammatory agents, transforming growth factor beta and heat shock. The increase in expression of HO-1 is thought to be a cellular defense mechanism against oxidative stress since elevated HO could eventually generate more bilirubin, an anti-oxidant.
TRUSTED_SUSTAINABILITY
Specifikationer
Specifikationer
HMOX1
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.09% sodium azide
P06762, P09601, P14901, Q5E9F2
HMOX1
Synthetic peptide corresponding to the sequence near the N-terminus (amino acids 1-30) of human Heme Oxygenase 1.
200 μg
Primary
Bovine, Canine, Human, Mouse, Rat
Antibody
IgG1 κ
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
HO-1-1
Unconjugated
HMOX1
bK286B10; D8Wsu38e; heat shock protein 32; heat shock protein, 32-kD; heme orygenase 1; heme oxygenase; heme oxygenase (decycling) 1; heme oxygenase (decyclizing) 1; heme oxygenase 1; heme oxygenase-1; Hemox; hemoxygenase; Heox; HEOXG; Hmox; Hmox1; HMOX1D; HO; Ho1; HO-1; Hsp32; P32 protein
Mouse
Protein G
RUO
15368, 24451, 3162, 442987, 513221
-20°C, Avoid Freeze/Thaw Cycles
Liquid