Läs mer
Abeba™ Business Men 32430 Shoes
Produktkod. 15669677
Change view
Klicka för att se tillgängliga alternativ
Storlek:
40
41
42
43
44
45
46
47
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Storlek
15669677 44
15629677 40
15639677 41
15649677 42
15659677 43
15679677 45
15689677 46
15699677 47
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 15669677 Leverantör Abeba™ Leverantörsnummer 32430/44
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Denna artikel kan inte returneras. Se returpolicy
ESD dissipative with high wearing comfort. Smooth leather construction.
  • CE EN ISO 20347:2012 O1 SRA
  • Smooth leather
  • Black
  • Slip resistance special rubber outsole sole
  • ESD dissipative according to DIN EN 61340
  • Lining microfiber, respiration absorbing
  • Instep area with lacing
  • Insole leather
Specifikationer
Certifikat och efterlevnad CE EN ISO 20347:2012 O1 SRA
Färg Black
Liner material Microfiber
Material Leather
Storlek 44
Enda funktioner Slip-resistant
Sulmaterial Rubber
Stil 32430