Läs mer
Brady™ BradyJet J2000 Colour Label Printer - EU with Brady Workstation LAB Suite
Produktkod. 16140501
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16140501
Brand: Brady™ 199966
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Denna artikel kan inte returneras. Se returpolicy
The BradyJet J2000 Colour Label Printer is a practical and easy to use, full colour inkjet label printer that can print reliable, high-resolution signs, facility and laboratory sample labels.
The perfect combo. Get the impact of colour—that’s wildly durable—to create a safer, more efficient workplace. Rely on the BradyJet J2000 Colour Label Printer when durable colour matters most.
  • Works great in a multitude of industries, from lab to safety and more
  • Compact design means it can snuggle in to tight spaces
  • On-demand, mid-volume inkjet solution creates photo-quality colour labels in a single pass
  • More colour options than you’d ever need—16 million!
  • Custom identification without the high cost of outsourced printing
  • Includes Brady Workstation Lab Suite to your solution and make label creation even easier
  • Setup is stress free – Contact our Brady Technical Support Team and we will schedule a free phone setup appointment
Specifikationer
Visningstyp None
Tryckteknik Inkjet
Mått (L x B) 389 x 264.00 mm
Färgkapacitet Multi-color
Inkluderar This kit contains a J2000 printer, a power cord and AC adapter, a USB cable, a Quick Start Guide, an ink cartridge, a material roll for printhead alignment, a USB stick with drivers and manuals and a Brady Workstation Laboratory Suite
Utskriftshastighet 63.50 mm/sec.
Upplösning 4800 dpi
Strömförsörjningsdetaljer 100/240 V