Läs mer
Novus Biologicals™ Recombinant Human ART5 GST (N-Term) Protein
Handla allt Bio Techne Produkter
Klicka för att se tillgängliga alternativ
Kvantitet:
10 μg
25 μg
Beskrivning
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Specifikationer
Specifikationer
| För användning med (applikation) | Western Blot, ELISA, Protein Array, Immunoaffinity Purification |
| Formulering | 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. |
| Gen-ID (Entrez) | 116969 |
| Molekylvikt (g/mol) | MolecularWeight-theroretical: 58.6 kDa |
| Kvantitet | 10 μg |
| Förvaringskrav | Store at -80°C. Avoid freeze/thaw cycles. |
| Gene Alias | ADP-ribosyltransferase 5, EC 2.4.2.31, ecto-ADP-ribosyltransferase 5, MGC22848, Mono(ADP-ribosyl)transferase 5, NAD(P)(+)--arginine ADP-ribosyltransferase 5 |
| Gensymbol | ART5 |
| Renhet eller kvalitetsklass | >80% by SDS-PAGE and Coomassie blue staining |
| Protein | ART5 |