Beskrivning
SSR2 Monoclonal antibody specifically detects SSR2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifikationer
Specifikationer
| Antigen | SSR2 |
| Användningsområden | Western Blot, ELISA, Immunofluorescence |
| Klassificering | Monoclonal |
| Klona | 31G6 |
| Konjugera | DyLight 594 |
| Formulering | 50 mM Sodium Borate |
| Gene Alias | DKFZp686F19123, Signal sequence receptor subunit beta, signal sequence receptor, beta (translocon-associated protein beta), SSR-beta, TLAP, translocon-associated protein beta, translocon-associated protein subunit beta, TRAP-beta, TRAPBTRAP-BETA |
| Värd art | Mouse |
| Immunogen | Recombinant human SSR2 (18-149aa) purified from E. coli |
| Reningsmetod | Protein A purified |
| Visa mer |