Läs mer
R&D Systems™ Recombinant Mouse ASIP/Agouti Protein
Klicka för att se tillgängliga alternativ
Kvantitet:
50 μg
Beskrivning
The Recombinant Mouse ASIP/Agouti Protein is derived from NS0. The Recombinant Mouse ASIP/Agouti Protein has been validated for the following applications: Bioactivity.
Specifikationer
Specifikationer
| Tillträdesnummer | Q03288 |
| Karakterisering | Bioactivity |
| För användning med (applikation) | Bioactivity |
| Formulering | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gen-ID (Entrez) | 434 |
| Molekylvikt (g/mol) | 12kDa |
| Kvantitet | 50 μg |
| Källa | Mouse myeloma cell line,NS0-derived mouse ASIP protein His23-Cys131 |
| Förvaringskrav | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Endotoxinkoncentration | <0.10EU per 1μg of the protein by the LAL method. |
| Visa mer |