Läs mer
ViewRNA™ ISH Cell Assay Kit
Specifikationer
Specifikationer
| Innehåll och lagring | Components sufficient for 1-plex to 3-plex (AlexaFluor 488, 546 and 647) assay in 24- well plate format (24 samples), 96 assays when using cover slips mounted on glass slides |
| Kvantitet | 1 kit |
| Typ | Detection Assay Kit |