missing translation for 'onlineSavingsMsg'
Learn More

Viking Termometerfabriken™Grid Dividers

Brand:  Tenak TE22736

Additional Details : Weight : 1.10000kg

Product Code. 15290444

  • 990.00 SEK / Pack of 36
Estimated Shipment: 19-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Cardboard
Grid Divider
100 and 130 mm Boxes
36 Pack
Square
For 100 and 130mm Boxes
7 (7 Cells of Less Than 17.75 mm O.D. Vials)
65 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers