Läs mer
Vilazodone Hydrochloride, MedChemExpress
Produktkod. 16486008
Encompass_Preferred
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16486008 10 mg
16402157 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16486008 Leverantör Medchem Express Leverantörsnummer HY14261
MedChemExpress Vilazodone Hydrochloride (EMD 68843 Hydrochloride) is a serotonin transporter (SER) inhibitor and 5-HT1A receptor partial agonist.
Vilazodone Hydrochloride (EMD 68843 Hydrochloride) is a serotonin transporter (SER) inhibitor and 5-HT1A receptor partial agonist.
Specifikationer
Kemiskt namn eller material Vilazodone Hydrochloride
Fysisk form Solid
CAS 163521-08-2
Färg Off-White
Kvantitet 10 mg
Anteckningar om renhetsgrad Research
Molekylformel C26H28ClN5O2
För användning med (applikation) Neuroscience-Neuromodulation
Synonym EMD 68843 HydrochlorideSB659746A Hydrochloride
Löslighetsinformation DMSO : 25 mg/mL (52.30 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
LEDER NC(C1=CC2=C(C=CC(N3CCN(CCCCC4=CNC5=CC=C(C=C54)C#N)CC3)=C2)O1)=O.[H]Cl
Molekylvikt (g/mol) 477.99
Formel vikt 477.99
Procent renhet 98.97%