missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Villin 1 Antibody (rVIL1/1325), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79943AF700
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Villin 1 Monoclonal specifically detects Villin 1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifikationer
| Villin 1 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate | |
| VIL1 | |
| A recombinant fragment (around aa179-311) of human Villin 1 protein (exact sequence is proprietary) (Uniprot: P09327) | |
| 0.1 mL | |
| Cancer, Cell Biology, Cellular Markers | |
| 7429 | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| rVIL1/1325 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| D2S1471, VIL, villin 1, villin-1 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Recognizes a protein of 95kDa, which is identified as villin. It is a major constituent in the microvilli, which compose the brush border of epithelial cells forming absorptive surfaces of the intestinal and renal proximal tubular epithelia. Anti-Villin labels the brush border area in the gastrointestinal mucosal epithelium and urogenital tract. Among neoplasms, villin is predominantly expressed in tumors of colorectal origin. Antibody to villin is useful in identifying malignant cells from primary and metastatic colorectal carcinomas. This antibody also labels Merkel cells of the skin. | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering