missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ Vinculin Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584619
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Immunogen sequence: SALTSKLADL RRQGKGDSPE ARALAKQVAT ALQNLQTKTN RAVANSRPAK AAVHLEGKIE QAQRWIDNPT VDDRGVGQAA IRGLVAEGHR LANVMMGP Highest antigen sequence indentity to the following orthologs: Mouse - 98%, Rat - 98%.
Vinculin (VCL) is a cytoskeletal protein associated with cell-cell and cell-extracellular matrix adherens-type junctions. It functions as one of several interacting proteins involved in anchoring F-actin to the membrane. It has been shown that a sequence of molecular interactions might be involved in the transmembrane assembly of adhesion plaques. In the assembly of adhesion plaques, the beta subunit of integrin binds to talin. Talin binds to vinculin that interacts with alpha-actinin and possibly with itself. Since alpha-actinin binds to and cross-links actin filaments, vinculin represents a key element in the transmembrane linkage of the extracellular matrix to the cytoplasmic microfilament system. Mutations in the gene can result in cardiomyopathy dilated 1W or familial hypertrophic 15.
Specifikationer
| Vinculin | |
| Polyclonal | |
| Unconjugated | |
| VCL | |
| 9430097D22; AA571387; AI462105; AW545629; CMD1W; CMH15; epididymis luminal protein 114; HEL114; metavinculin; meta-vinculin; metavinculin {ECO:0000250; MV; MVCL; RP11-178G16.3; UniProtKB:P18206}; unnamed protein product; Vcl; vcl {ECO:0000250; VIN; vinc; vinc 1; VINC1; Vinculin; vinculin {ECO:0000250 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7414 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P18206 | |
| VCL | |
| Recombinant protein corresponding to Human VCL. Recombinant protein control fragment (Product #RP-105286). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering