Läs mer
Invitrogen™ Vinculin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595919
Beskrivning
Positive Samples: 293T, NCI-H460, HepG2, BxPC-3, U-87MG, Mouse heart, Mouse brain; Cellular Location: Cell junction, Cell membrane, Cytoplasm, Cytoplasmic side, Peripheral membrane protein, adherens junction, cytoskeleton, focal adhesion Immunogen sequence: AKREVENSED PKFREAVKAA SDELSKTISP MVMDAKAVAG NISDPGLQKS FLDSGYRILG AVAKVREAFQ PQEPDFPPPP PDLEQLRLTD ELAPPKPPLP EGEVPPPRPP PPEEKDEEFP EQKAGEVINQ PMMMAARQLH DEARKWSSKG NDIIAAAKRM ALLMAEMSRL VRGGSGTKRA LIQCAKDIAK ASDEVTRLAK EVAKQCTDKR IRTNLLQVCE RIPTISTQLK ILSTVKATML GRTNISDEES EQATEMLVHN AQNLMQSVKE TVREAEAASI KIRTDAGFTL RWVRKTPWYQ.
Vinculin (VCL) is a cytoskeletal protein associated with cell-cell and cell-extracellular matrix adherens-type junctions. It functions as one of several interacting proteins involved in anchoring F-actin to the membrane. It has been shown that a sequence of molecular interactions might be involved in the transmembrane assembly of adhesion plaques. In the assembly of adhesion plaques, the beta subunit of integrin binds to talin. Talin binds to vinculin that interacts with alpha-actinin and possibly with itself. Since alpha-actinin binds to and cross-links actin filaments, vinculin represents a key element in the transmembrane linkage of the extracellular matrix to the cytoplasmic microfilament system. Mutations in the gene can result in cardiomyopathy dilated 1W or familial hypertrophic 15.
Specifikationer
| Vinculin | |
| Polyclonal | |
| Unconjugated | |
| VCL | |
| 9430097D22; AA571387; AI462105; AW545629; CMD1W; CMH15; epididymis luminal protein 114; HEL114; metavinculin; meta-vinculin; metavinculin {ECO:0000250; MV; MVCL; RP11-178G16.3; UniProtKB:P18206}; unnamed protein product; Vcl; vcl {ECO:0000250; VIN; vinc; vinc 1; VINC1; Vinculin; vinculin {ECO:0000250 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 22330, 305679, 7414 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.69 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P18206, P85972, Q64727 | |
| VCL | |
| Synthetic peptide. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.