Läs mer
Invitrogen™ Vinculin Recombinant Rabbit Monoclonal Antibody (9I3Y8)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA542795
Beskrivning
Positive test controls include: HeLa, HepG2, RD, Mouse liver, Mouse brain, Rat liver, Rat skeletal muscle, Rat brain. The target is usually found in the following locations: Cell junction, Cell membrane, Cytoplasm, Cytoplasmic side, Peripheral membrane protein, adherens junction, cytoskeleton, focal adhesion. Immunogen sequence: DAKAVAGNIS DPGLQKSFLD SGYRILGAVA KVREAFQPQE PDFPPPPPDL EQLRLTDELA PPKPPLPEGE VPPPRPPPPE EKDEEFPEQK AGEVINQPMM M.
Vinculin (VCL) is a cytoskeletal protein associated with cell-cell and cell-extracellular matrix adherens-type junctions. It functions as one of several interacting proteins involved in anchoring F-actin to the membrane. It has been shown that a sequence of molecular interactions might be involved in the transmembrane assembly of adhesion plaques. In the assembly of adhesion plaques, the beta subunit of integrin binds to talin. Talin binds to vinculin that interacts with alpha-actinin and possibly with itself. Since alpha-actinin binds to and cross-links actin filaments, vinculin represents a key element in the transmembrane linkage of the extracellular matrix to the cytoplasmic microfilament system. Mutations in the gene can result in cardiomyopathy dilated 1W or familial hypertrophic 15.
Specifikationer
| Vinculin | |
| Recombinant Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| P18206, P85972, Q64727 | |
| VCL | |
| Synthetic peptide. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 9I3Y8 | |
| Unconjugated | |
| VCL | |
| 9430097D22; AA571387; AI462105; AW545629; CMD1W; CMH15; epididymis luminal protein 114; HEL114; metavinculin; meta-vinculin; metavinculin {ECO:0000250; MV; MVCL; RP11-178G16.3; UniProtKB:P18206}; unnamed protein product; Vcl; vcl {ECO:0000250; VIN; vinc; vinc 1; VINC1; Vinculin; vinculin {ECO:0000250 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 22330, 305679, 7414 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.