Läs mer
Vindesine Sulfate Salt, TRC
Produktkod. 30457186
Change view
Kvantitet:
2.5 mg
10 mg
25 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30457186 10 mg -
30452294 25 mg -
30450362 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30457186 Leverantör Toronto Research Chemicals Leverantörsnummer TRCV41455010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 59917-39-4
Molekylformel C43 H55 N5 O7 . H2 O4 S
Molekylvikt (g/mol) 852
Synonym Vindesine sulfate, Methyl (5S,7R,9S)-9-[(3aR,4R,5S,5aR,10bR,13aR)-5-carbamoyl-3a-ethyl-4,5-dihydroxy-8-methoxy-6-methyl-3a,4,5,5a,6,11,12,13a-octahydro-1H-indolizino[8,1-cd]carbazol-9-yl]-5-ethyl-5-hydroxy-1,4,5,6,7,8,9,10-octahydro-2H-3,7-methanoazacycloundecino[4,5-b]indole-9-carboxylate sulfate, Vincaleukoblastine, 3-(aminocarbonyl)-O4-deacetyl-3-de(methoxycarbonyl)-, sulfate (1:1), Vincaleukoblastine, 3-(aminocarbonyl)-O4-deacetyl-3-de(methoxycarbonyl)-, sulfate (1:1) (salt) (9CI), 1H-Indolizino[8,1-cd]carbazole, vincaleukoblastine deriv., 2H-3,7-Methanoazacycloundecino[5,4-b]indole, vincaleukoblastine deriv., DAVA, Desacetylvinblastine amide sulfate, Eldesine, Eldisine, Fildesin, LY 099094, Lilly 99094, NSC 245467, Vindesine sulfate, Xi Ai Ke Vial
LEDER CC[C@]1(O)C[C@@H]2CN(CCc3c([nH]c4ccccc34)[C@@](C2)(C(=O)OC)c5cc6c(cc5OC)N(C)[C@H]7[C@](O)([C@H](O)[C@]8(CC)C=CCN9CC[C@]67[C@H]89)C(=O)N)C1.OS(=O)(=O)O
Specifikationer
Kemiskt namn eller material Vindesine Sulfate
Kvantitet 10 mg
InChI formel InChI=1S/C43H55N5O7.H2O4S/c1-6-39(52)21-25-22-42(38(51)55-5,33-27(13-17-47(23-25)24-39)26-11-8-9-12-30(26)45-33)29-19-28-31(20-32(29)54-4)46(3)35-41(28)15-18-48-16-10-14-40(7-2,34(41)48)36(49)43(35,53)37(44)50;1-5(2,3)4/h8-12,14,19-20,25,34-36,45,49,52-53H,6-7,13,15-18,21-24H2,1-5H3,(H2,44,50);(H2,1,2,3,4)/t25-,34-,35+,36+,39-,40+,41+,42-,43-;/m0./s1
Formel vikt 851.3775