Läs mer
Vinorelbine N'b-Oxide, TRC
Produktkod. 30450114
Change view
Kvantitet:
2.5 mg
25 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30450114 2.5 mg 2.5mg
30450208 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30450114 Leverantör Toronto Research Chemicals Leverantörsnummer TRCV3151002.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 74075-34-6
Molekylformel C45H54N4O9
Molekylvikt (g/mol) 794.93
Synonym (2beta,3beta,4beta,5alpha,12R,19alpha)-4-(Acetyloxy)-6,7-didehydro-15-[(2R,6R,8S)-4-ethyl-1,3,6,7,8,9-hexahydro-8-(methoxycarbonyl)-2-oxido-2,6-methano-2H-azecino[4,3-b]indol-8-yl]-3-hydroxy-16-methoxy-1-methyl-Aspidospermidine-3-carboxylic Acid Methyl Ester,
LEDER C(C)[C@@]12[C@]3([C@@]4([C@]([C@](C(OC)=O)(O)[C@@H]1OC(C)=O)(N(C)C=5C4=CC(=C(OC)C5)[C@]6(C(OC)=O)C7=C(C=8C(N7)=CC=CC8)CN9(=O)C[C@](C6)(C=C(CC)C9)[H])[H])CCN3CC=C2)[H]
Specifikationer
Kemiskt namn eller material Vinorelbine N'b-Oxide
Kvantitet 2.5 mg
InChI formel InChI=1S/C45H52N4O9/c1-8-27-19-28-22-44(40(51)56-6,36-30(25-49(54,23-27)24-28)29-13-10-11-14-33(29)46-36)32-20-31-34(21-35(32)55-5)47(4)38-43(31)16-18-48-17-12-15-42(9-2,37(43)48)39(58-26(3)50)45(38,53)41(52)57-7/h10-15,19-21,25,28,37-39,53H,8-9,16-18,22-24H2,1-7H3/p+1/t28?,37-,38+,39+,42+,43+,44+,45-,49+/m0/s1
Formel vikt 794.3891