missing translation for 'onlineSavingsMsg'
Learn More

Azpack™ Vinyl Labeling Tape

Label the contents of any item easily. Azpack™ Vinyl Labeling Tape is great for writing on and adheres to most surfaces.

Marque:  Azpack™ LAB-890-T

Informations supplémentaires : Weight : 0.09400kg

Code produit. 12365758

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Voir les produits de remplacement »


Description

Description

  • Opaque cellulose surface
  • Ideal for marking and labeling
  • Self-adhesive
Spécification

Spécification

Label Tape
Cellulose
25 mm
Opaque
1 Ea.
White
66 m
66 m x 25 mm
Roll
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions