Learn More
Brady™ Vinyl Coated High Tensile Steel Safety Lockout
Make sure all machinery stays locked until every individual working on the repairs or inspection is finished.
Brand:  Brady™ 805842
Additional Details : Weight : 0.09300kg
Product Code. 15673537
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Each person puts a lock on the hasp, but the hasp cannot be removed until all lockouts have been removed, thus preventing accidents.
Specifications
Specifications
Safety Lockout
Vinyl-coated High Tensile Steel
58.42 x 129.29 mm
1 Ea.
25.00 mm
Yellow
Safety Lockout
129.29 mm
58.42 mm