Learn More
Vinylbenzyl Chloride, Mixture of 2, 3- and 4-isomers (Stabilized with TBC), TRC
Product Code. 30456744
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456744 1 g 1g
30449882 10 g 10g
30449068 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30456744 Supplier Toronto Research Chemicals Supplier No. TRCV7570531G
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 30030-25-2
Molecular Formula 2 C9 H9 Cl
Molecular Weight (g/mol) 305.241
Synonym (Chloromethyl)ethenylbenzene, (Chloromethyl)styrene, CMS-AM, CMS-P, ar-(Chloromethyl)styrene, ar-Vinylbenzyl chloride
IUPAC Name 1-(chloromethyl)-3-ethenylbenzene;1-(chloromethyl)-4-ethenylbenzene
SMILES ClCc1ccc(C=C)cc1.ClCc2cccc(C=C)c2
Specifications
Quantity 1 g
InChI Formula InChI=1S/2C9H9Cl/c1-2-8-3-5-9(7-10)6-4-8;1-2-8-4-3-5-9(6-8)7-10/h2*2-6H,1,7H2
Formula Weight 304.079
Chemical Name or Material Vinylbenzyl Chloride, Mixture of 2, 3- and 4-isomers