Läs mer
Viral Dengue Virus 4 NS1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB9444-100
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Dengue Virus 4 NS1 Monoclonal specifically detects Dengue Virus 4 NS1 in Viral samples. It is validated for Western Blot.
Specifikationer
| Dengue virus 4 NS1 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1 ug/mL | |
| NP_740318 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Detects recombinant Dengue Virus 4 NS1 in direct and capture ELISAs and Western blots. In direct ELISAs and Western Blots, no cross-reactivity with recombinant Dengue Virus 1, 2 and 3 NS1 proteins was observed. | |
| Virus | |
| Purified |
| Western Blot | |
| 971314 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DEN-4, Dengue NS1, Dengue pan, Dengue virus Serotype 4, Dengue virus Type 4, DENV4, DENV4-NS1 | |
| Human embryonic kidney cell line HEK293-derived recombinant Dengue Virus 4 NS1 protein Asp1-Ala352 Accession # NP_740318 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |