Läs mer
Ansell™ Virtex™ 79-700 Series Blue Nitrile Immersion Gloves
Handla allt Ansell Produkter
Storlek:
7
8
9
10
11
Förpackningsstorlek:
50 par
Beskrivning
- Reusable
- Anti-static according to EN1149
- With Aquadri™-Ansell Moisture Management Technology™, which gives excellent moisture absorption capabilities and keeps users' hands drier for longer
- Significantly lower tendency to shed than traditional cotton flock gloves
- Ideal for light assembly in dry, fully wet or oily conditions, engine and system testing, paint and spray shops, food distribution
Specifikationer
Specifikationer
| Material på baksidan | Nitrile |
| Manschettmaterial | Nitrile |
| Produkttyp | Immersion Gloves |
| Storlek | 11 |
| Stil | 79-700 |
| Tjocklek (metrisk) | 0.2 mm |
| Färg på handskemanschett | Blue |
| Handske manschettstil | Straight |
| Liner material | Aquadri |
| Kvantitet | 1 Pair |
| Visa mer |