Learn More
Thermo Scientific™ VISIONlife™ UV-Visible Spectrophotometer Software
Thermo Scientific™ VISIONlife is an optional module to perform sophisticated kinetics assay with user-definable equations and nucliec acid and protein denturation/renaturation assays.
Brand: Thermo Scientific™ 10040201
Additional Details : Weight : 0.10000kg
Description
Thermo Scientific VISIONlife is an optional module to perform sophisticated kinetics assay with user-definable equations and nucliec acid and protein denaturation/renaturation assays.
Powerful, versatile software offers advanced instrument control and data manipulation capabilities. Optimized for Thermo Scientific UV-Visible spectrophotometers, it brings a user-friendly, graphical environment to spectrophotometric analysis. Method setup, data collection, calculations, report presentation, regulatory compliance - all aspects of your analysis are easier with this software.
- For use with Evolution™ 100, 160, 300 and 600 UV-Vis Spectrophotometers
- Use with VISIONpro™ or VISIONsecurity™ software
- Full-featured Rate application provides serial, parallel, and triggered rate modes
- Integrated Kinetic Analysis software allows modeling of rate and concentration data for enzyme behavior, from basic Michaelis-Menten to binding models including Hill plots, or user-entered equations
- DNA melting application enables advanced control of the heating/cooling cycles—saving valuable time and allowing experiments to be performed up to six times faster than with traditional fixed temperature ramps
- Wide range of Tm calculation methods included
Includes:
User Manual
Requires:
System Requirements: Microsoft Windows™ 2000, XP, Vista; plus VISIONpro (Part No. 10040101, 10040121) or VISIONsecurity (10040301, 10040341)
Specifications
FDA guidelnes, IEEE software engineering standards | |
Life Science | |
Pre-programmed Nucleic Acid and Protein Methods, Rate, DNA Melting, Enzyme Kinetics |
PC Software | |
Evolution 300 | |
At least Windows XP, plus VISIONpro (Part No. 10040101, 10040121) or VISIONsecurity (10040301, 10040341) |