Learn More
Macherey-Nagel™ Visocolor™ ECO Chromium (VI) Colorimetric Test Kit
Brand: Macherey-Nagel™ 931020
Additional Details : Weight : 0.07250kg Transport : UN number : 3316 Chem class : 9 Pack group : II
Specifications
Chromium compounds are present in trivalent form [chromium (III) ions] and in hexavalent form (chromate and dichromate ions) in industrial waste water | |
0.02 to 0.5mg / l Cr (VI) | |
ECO Chromium (VI) Colorimetric Test Kit |
1 Each | |
140 Provisions |
Safety and Handling
- VISOCOLOR ECO Chromium (VI)
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 B
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : o-phosphoric acid, dimethyl sulfoxide